missing translation for 'onlineSavingsMsg'
Learn More

SLC38A4 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18415888 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415888 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415888 Supplier Novus Biologicals Supplier No. NBP281807FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC38A4 Polyclonal antibody specifically detects SLC38A4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Amino acid transporter A3, ATA3MGC126876, FLJ10191, N amino acid transporter 3, Na(+)-coupled neutral amino acid transporter 4, NAT3amino acid transporter system A3, PAAT, SNAT4, sodium-coupled neutral amino acid transporter 4, Solute carrier family 38 member 4, solute carrier family 38, member 4, System A amino acid transporter 3, System N amino acid transporter 3
Host Species Rabbit
Immunogen SLC38A4 antibody was raised against an 18 amino acid peptide from near the center of human SLC38A4. The immunogen is located within amino acids 270 - 320 of SLC38A4. Amino Acid Squence: MDYTHRNPAGLDENQAKG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55089
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.