missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC38A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549591
This item is not returnable.
View return policy
Description
SLC38A3 Polyclonal Antibody for Western Blot, Flow
As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.
Specifications
| SLC38A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A3 | |
| 0610012J02Rik; D9Ucla2; G17; mNAT; N system amino acids transporter NAT-1; Na(+)-coupled neutral amino acid transporter 3; Na+ and H+ coupled amino acid transport system N1; NAT1; N-system amino acid transporter 1; RGD:628620}; Slc38-3; Slc38a3; slc38a3 {ECO:0000312; SLC38A3 protein; Sn1; Snat3; sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; solute carrier family 38, member 3; system N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q99624 | |
| SLC38A3 | |
| KLH conjugated synthetic peptide between 442-468 amino acids from the Central region of human SLC38A3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction