missing translation for 'onlineSavingsMsg'
Learn More

SLC38A3 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18364563
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364563 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364563 Supplier Bio-Techne Supplier No. NBP306519G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC38A3 Polyclonal antibody specifically detects SLC38A3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A3
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias G17sodium-coupled neutral amino acid transporter 3, Na(+)-coupled neutral amino acid transporter 3, NAT1, N-system amino acid transporter 1, SN1system N1 Na+ and H+-coupled glutamine transporter, SNAT3, Solute carrier family 38 member 3, solute carrier family 38, member 3, System N amino acid transporter 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC38A3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10991
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.