missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC38A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557187
This item is not returnable.
View return policy
Description
Immunogen sequence: MKKAEMGRFS ISPDEDSSSY SSNSDFNYSY PTKQAALKSH YADVDPENQN FLLESNLGKK KYETEFHP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.
Under hypertonic conditions the induction of SLC38A2/SNAT2 leads to the stimulation of transport system A and to the increase in the cell content of amino acids. Its amino acid response element, along with a nearby conserved CAAT box, has enhancer activity in that it functions in an orientation and position independent manner, and it confers regulated transcription to a heterologous promoter.
Specifications
| SLC38A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A2 | |
| 5033402L14Rik; AI316867; amino acid system A transporter; amino acid transporter 2; amino acid transporter A2; amino acid transporter cationic 2; amino acid transporter system A2; amino acid transporter, cationic 2 (low affinity); ATA2; Atrc2; Kiaa1382; mKIAA1382; PRO1068; Protein 40-9-1; Sa1; Sat2; SLC38A2; Snat2; sodium-coupled neutral amino acid transporter 2; Solute carrier family 38 member 2; solute carrier family 38, member 2; system A amino acid transporter 2; System A transporter 1; system N amino acid transporter 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54407 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QD8 | |
| SLC38A2 | |
| Recombinant protein corresponding to Human SLC38A2. Recombinant protein control fragment (Product #RP-95791). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction