missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A1 Antibody (S104-32), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59311-25ug
This item is not returnable.
View return policy
Description
SLC38A1 Monoclonal specifically detects SLC38A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SLC38A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| SLC38A1 | |
| Fusion protein amino acids 1- 63 of rat SNAT1 | |
| 25 μg | |
| Primary | |
| Detects 50kDa. | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| S104-32 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 81539 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction