missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A1 Antibody (S104-32), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59311F
This item is not returnable.
View return policy
Description
SLC38A1 Monoclonal specifically detects SLC38A1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SLC38A1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SLC38A1 | |
| Fusion protein amino acids 1- 63 of rat SNAT1 | |
| 100 μL | |
| 81539 | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S104-32 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects 50kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction