missing translation for 'onlineSavingsMsg'
Learn More

SLC38A1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18379821 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379821 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379821 Supplier Novus Biologicals Supplier No. NBP306520AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC38A1 Polyclonal antibody specifically detects SLC38A1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC38A1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81539
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.