missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC37A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558599
This item is not returnable.
View return policy
Description
Immunogen sequence: LDKDDLGFIT SSQSAAYAIS KFVSGVLSDQ MS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
Specifications
| SLC37A4 | |
| Polyclonal | |
| Unconjugated | |
| Slc37a4 | |
| G6PT; G6PT1; G6PT2; G6PT3; glucose 6-phosphate transporter; glucose-5-phosphate transporter; glucose-6-phosphatase transport protein 1; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; glucose-6-phosphatase, transport protein 1; glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; GSD1b; GSD-1b; GSD1c; GSD1d; mG6PT; MGC15729; microsomal glucose-6-phosphate transporter; PRO0685; SLC37A4; solute carrier family 37 (glucose-6-phosphate transporter), member 4; solute carrier family 37 (glycerol-6-phosphate transporter), member 4; solute carrier family 37 member 4; Transformation-related gene 19 protein; transporter; TRG19; TRG-19; Unknown (protein for MGC:137413) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2542 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43826 | |
| Slc37a4 | |
| Recombinant protein corresponding to Human SLC37A4. Recombinant protein control fragment (Product #RP-95793). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction