missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC37A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542500
This item is not returnable.
View return policy
Description
Peptide sequence: FFGLLVSPEE IGLSGIEAEE NFEEDSHRPL INGGENEDEY EPNYSIQDDS Sequence homology: Cow: 92%; Dog: 86%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Rabbit: 93%; Rat: 85%.
SLC37A3 (Solute Carrier Family 37 Member 3) is a Protein Coding gene. Diseases associated with SLC37A3 include Glycerol Kinase Deficiency and Deafness, Autosomal Recessive 8. Gene Ontology (GO) annotations related to this gene include transporter activity and transmembrane transporter activity. An important paralog of this gene is SLC37A1. [GeneCards].
Specifications
| SLC37A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC37A3 | |
| SLC37A3; solute carrier family 37 (glycerol-3-phosphate transporter), member 3; solute carrier family 37 member 3; solute carrier family 37, member 3; SPX3; Sugar phosphate exchanger 3; sugar phosphate exchanger 3-like protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84255 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NCC5 | |
| SLC37A3 | |
| Synthetic peptide directed towards the middle region of human SLC37A3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction