missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC37A2 Polyclonal Antibody

SLC37A2 Polyclonal Antibody
Brand: Invitrogen PA5107265
This item is not returnable.
View return policy
Description
SLC37A2 Polyclonal Antibody for Western Blot
Specifications
| SLC37A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC37A2 | |
| cAMP inducible 2 protein; cAMP inducible gene 2; cAMP-inducible protein 2; ci2; cI-2; G3PP; glucose-6-phosphate exchanger SLC37A2; pp11662; RGD1564160; Slc37a1; SLC37A2; solute carrier family 37 (glucose-6-phosphate transporter), member 2; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 2; solute carrier family 37 member 2; sugar phosphate exchanger 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9WU81 | |
| SLC37A2 | |
| Synthetic peptide directed towards the middle region of Mouse Slc37a2 (aa 300-350). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction