missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC37A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556468
This item is not returnable.
View return policy
Description
Immunogen sequence: IVKGELHKYC TAWDEADVRF SSQNRKSGSA APHQLPDNET DCGWAPFDKN NY Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 55%.
SLC37A1, a member of the sugar-phosphate transport family, transports glycerol-3-phosphate (G3P) between cellular compartments for its utilization in several compartment-specific biochemical pathways.
Specifications
| SLC37A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC37A1 | |
| FLJ22340; G-3-P permease; G-3-P transporter; G3PP; Glucose-6-phosphate exchanger SLC37A1; glycerol-3-phosphate permease; glycerol-3-phosphate transporter; Slc37a1; solute carrier family 37 (glucose-6-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; solute carrier family 37 member 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57057 | |
| SLC37A1 | |
| Recombinant protein corresponding to Human SLC37A1. Recombinant protein control fragment (Product #RP-95891). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction