missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC36A4 Antibody (PAT-4/9/H10), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50237AF700
This item is not returnable.
View return policy
Description
SLC36A4 Monoclonal specifically detects SLC36A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockdown Validated.
Specifications
| SLC36A4 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SLC36A4 | |
| Antigenic amino acid sequence within the N-terminus of SLC36A4 (REELDMDVMRPLINE-C). | |
| 0.1 mL | |
| Cellular Signaling, metabolism | |
| 120103 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, KnockDown | |
| PAT-4/9/H10 | |
| Western Blot, Immunohistochemistry | |
| proton/amino acid transporter 4, solute carrier family 36 (proton/amino acid symporter), member 4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction