missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC36A4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98630CL1
This item is not returnable.
View return policy
Description
SLC36A4 Polyclonal antibody specifically detects SLC36A4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| SLC36A4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| proton/amino acid transporter 4, solute carrier family 36 (proton/amino acid symporter), member 4 | |
| Produced in rabbits immunized with E. coli-derived Human SLC36A4 fragment. | |
| 0.1 mL | |
| Cellular Signaling, metabolism | |
| 120103 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction