Learn More
SLC36A3 Mouse anti-Human, Unconjugated, Clone: 1A8E6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67929-1-IG-20UL
This item is not returnable.
View return policy
Description
The SLC36A3 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human SLC36A3. This antibody recognizes human, mouse antigen. The SLC36A3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SLC36A3 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 285641 | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 1A8E6 | |
| Mouse | |
| Primary | |
| Human, Mouse |