missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC36A2 Polyclonal Antibody
SLC36A2 Polyclonal Antibody
Brand: Thermo Scientific PA564009
This item is not returnable.
View return policy
Description
SLC36A2 Polyclonal Antibody for IHC
Specifications
| SLC36A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC36A2 | |
| FLJ16051; MGC119658; MGC119660; PAT2; Proton/amino acid transporter 2; Proton-coupled amino acid transporter 2; SLC36A2; solute carrier family 36 (proton/amino acid symporter), member 2; solute carrier family 36 member 2; TRAMD1; tramdorin-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 153201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q495M3 | |
| SLC36A2 | |
| Recombinant protein corresponding to Human SLC36A2. Recombinant protein control fragment (Product #RP-103873). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction