missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC36A2 Polyclonal antibody specifically detects SLC36A2 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Applications | ELISA |
| Gene Alias | FLJ16051, MGC119658, MGC119660, PAT2proton-coupled amino acid transporter 2, Proton/amino acid transporter 2, solute carrier family 36 (proton/amino acid symporter), member 2, Solute carrier family 36 member 2, TRAMD1tramdorin-1, tramdorin, Tramdorin-1 |
| Gene ID (Entrez) | 153201 |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?