missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35F5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542494
This item is not returnable.
View return policy
Description
Peptide sequence: VDREDKLDIP MFFGFVGLFN LLLLWPGFFL LHYTGFEDFE FPNKVVLMCI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
SLC35F5 is a putative solute transporter.
Specifications
| SLC35F5 | |
| Polyclonal | |
| Unconjugated | |
| SLC35F5 | |
| 1300003P13Rik; AI646727; HCV NS5A-transactivated protein 3; hepatitis C virus NS5A-transactivated protein 3; NS5ATP3; SLC35F5; Solute carrier family 35 member F5; solute carrier family 35, member F5; UNQ2545/PRO6097 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 80255 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WV83 | |
| SLC35F5 | |
| Synthetic peptide directed towards the C-terminal of human SLC35F5 (aa 351-400). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction