missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35F1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107023
This item is not returnable.
View return policy
Description
Sequence homology: Dog: 83%; Guinea Pig: 83%; Human: 100%; Mouse: 80%.
SLC35F1 is a putative solute transporter.
Specifications
| SLC35F1 | |
| Polyclonal | |
| Unconjugated | |
| SLC35F1 | |
| C6orf169; dJ230I3.1; SLC35F1; Solute carrier family 35 member F1; solute carrier family 35, member F1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 222553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q5T1Q4 | |
| SLC35F1 | |
| Synthetic peptide directed towards the N terminal region of human SLC35F1 (aa 1-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction