missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC35D3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA572721

Product Code. 15901385

  • £441.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SLC35D3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

May play a role in hemostasis as a regulator of the biosynthesis of platelet-dense granules.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC35D3
Polyclonal
Unconjugated
SLC35D3
6230421J19Rik; frc, fringe-like 1; Frcl1; Fringe connection-like protein 1; SLC35D3; Solute carrier family 35 member D3; solute carrier family 35, member D3
Rabbit
Antigen affinity chromatography
RUO
340146, 76157
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q5M8T2, Q8BGF8
SLC35D3
SLC35D3 antibody for a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is within the last 50 amino acids of SLC35D3.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.