missing translation for 'onlineSavingsMsg'
Learn More

SLC35D3 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18417536 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417536 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417536 Supplier Novus Biologicals Supplier No. NBP281839V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC35D3 Polyclonal antibody specifically detects SLC35D3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35D3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias bA55K22.3, frc, fringe-like 1, FRCL1, Fringe connection-like protein 1, MGC102873, solute carrier family 35 member D3, solute carrier family 35, member D3
Host Species Rabbit
Immunogen SLC35D3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is located within the last 50 amino acids of SLC35D3. Amino Acid Squence: LVRGTRYMKKDYLIENE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 340146
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.