missing translation for 'onlineSavingsMsg'
Learn More

SLC35D2 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18407917 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407917 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407917 Supplier Novus Biologicals Supplier No. NBP281806UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC35D2 Polyclonal antibody specifically detects SLC35D2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35D2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias HFRC, HFRC1UDP-N-acetylglucosamine transporter, Homolog of Fringe connection protein 1, MGC117215, MGC142139, Solute carrier family 35 member D2, solute carrier family 35, member D2, SQV7Lfringe connection, SQV7-like protein, UDP-galactose transporter-related protein 8, UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter, UGTREL8, UGTrel8hfrc
Host Species Rabbit
Immunogen SLC35D2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human SLC35D2. The immunogen is located within the last 50 amino acids of SLC35D2. Amino Acid Squence: SSQLKPKPVGEENI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11046
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.