missing translation for 'onlineSavingsMsg'
Learn More

SLC35D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18379766 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379766 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379766 Supplier Novus Biologicals Supplier No. NBP309954100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC35D1 Polyclonal specifically detects SLC35D1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35D1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias KIAA0260MGC138236, solute carrier family 35 (UDP-glucuronic acid/UDP-N-acetylgalactosamine dualtransporter), member D1, Solute carrier family 35 member D1, UDP-galactose transporter-related protein 7, UDP-GlcA/UDP-GalNAc transporter, UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter, UGTrel7, UGTREL7UDP-galactose transporter-related 7
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human S35D1. Peptide sequence GGDYIFTWTNFIGLNISIAGSLVYSYITFTEEQLSKQSEANNKLDIKGKG
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23169
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.