missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35C2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567820
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Human samples.
May play an important role in the cellular response to tissue hypoxia. May be either a GDP-fucose transporter that competes with SLC35C1 for GDP-fucose, or a factor that otherwise enhances the fucosylation of Notch and is required for optimal Notch signaling in mammalian cells (By similarity).Specifications
SLC35C2 | |
Polyclonal | |
Unconjugated | |
SLC35C2 | |
BA394O2.1, C20orf5, CGI-15, OVCOV1, ovarian cancer overexpressed 1, ovarian cancer-overexpressed gene 1 protein, solute carrier family 35 member C2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
228875, 51006 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NQQ7, Q8VCX2 | |
SLC35C2 | |
Synthesized peptide derived from human SLC35C2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |