missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35C2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83548-0.1ML
This item is not returnable.
View return policy
Description
SLC35C2 Polyclonal specifically detects SLC35C2 in Mouse samples. It is validated for Western Blot.
Specifications
| SLC35C2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51006 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| bA394O2.1, C20orf5, CGI-15, FLJ37039, FLJ46434, MGC20633, MGC32079, MGC39183, ovarian cancer overexpressed 1, Ovarian cancer-overexpressed gene 1 protein, OVCOV1, solute carrier family 35 member C2, solute carrier family 35, member C2 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of mouse SLC35C2 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction