missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC35C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29502
This item is not returnable.
View return policy
Description
SLC35C2 Polyclonal antibody specifically detects SLC35C2 in Human,Mouse samples. It is validated for Western Blot
Specifications
| SLC35C2 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51006 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| bA394O2.1, C20orf5, CGI-15, FLJ37039, FLJ46434, MGC20633, MGC32079, MGC39183, ovarian cancer overexpressed 1, Ovarian cancer-overexpressed gene 1 protein, OVCOV1, solute carrier family 35 member C2, solute carrier family 35, member C2 | |
| Between 315 and 365 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction