missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556220
This item is not returnable.
View return policy
Description
Immunogen sequence: FRRKNYLETG RGLCFPLVKA CVFGNEPKAS DEVPLAPRTE AAETT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
Mediates the transport of adenosine 3'-phospho 5'-phosphosulfate (PAPS), from cytosol into Golgi. PAPS is a universal sulfuryl donor for sulfation events that take place in the Golgi. May indirectly participate in activation of the NF-kappa-B and MAPK pathways.
Specifications
| SLC35B2 | |
| Polyclonal | |
| Unconjugated | |
| SLC35B2 | |
| 1110003M08Rik; 3'-phosphoadenosine 5'-phosphosulfate transporter; AA407792; Adenosine 3'-phospho 5'-phosphosulfate transporter 1; AI842275; J207; PAPS transporter 1; PAPST1; PSEC0149; Putative MAPK-activating protein PM15; putative NF-kappa-B-activating protein 48; Slc35b1; SLC35B2; SLL; solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2; solute carrier family 35 member B2; solute carrier family 35 member B2 variant 2; solute carrier family 35, member B1; solute carrier family 35, member B2; UGTrel4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 347734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TB61 | |
| SLC35B2 | |
| Recombinant protein corresponding to Human SLC35B2. Recombinant protein control fragment (Product #RP-94917). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction