missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £443.00
Specifications
| Antigen | SLC35B2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18467910
|
Novus Biologicals
NBP1-80641-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18288766
|
Novus Biologicals
NBP1-80641 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC35B2 Polyclonal specifically detects SLC35B2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SLC35B2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 347734 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FRRKNYLETGRGLCFPLVKACVFGNEPKASDEVPLAPRTEAAETT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| adenosine 3'-phospho 5'-phosphosulfate transporter 1, PAPS transporter 1, PAPST13'-phosphoadenosine 5'-phosphosulfate transporter, Putative MAPK-activating protein PM15, Putative NF-kappa-B-activating protein 48, SLL, Solute carrier family 35 member B2, solute carrier family 35 member B2 variant 2, solute carrier family 35, member B2, UGTrel4 | |
| SLC35B2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title