missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565523
This item is not returnable.
View return policy
Description
Immunogen sequence: QFVNYPTQVLG KSCKPIPVML LGVTLLKKKY P Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
SLC35B1 belongs to the nucleotide-sugar transporter family and it is probable sugar transporter.
Specifications
| SLC35B1 | |
| Polyclonal | |
| Unconjugated | |
| SLC35B1 | |
| Endoplasmic reticulum nucleotide sugar transporter 1; ERNST; galactose transporter; hUGTrel1; putative endoplasmic reticulum nucleotide sugar transporter; Slc35b1; Solute carrier family 35 member B1; solute carrier family 35 member B1-like protein; solute carrier family 35, member B1; UDP-galactose translocator 2; UDP-galactose transporter related; UDP-galactose transporter-related protein 1; Ugalt2; UGALT2 protein; UGTREL1; wu:fj99g11; zgc:92284 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10237 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78383 | |
| SLC35B1 | |
| Recombinant Human SLC35B1. Recombinant protein control fragment (Product #RP-100118). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction