missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566117
This item is not returnable.
View return policy
Description
Immunogen sequence: KPQVPEYAPRQ ERIRDLSGNL WERSSGDGEE LERLTKPKSD ESDEDTF Highest antigen sequence identity to the following orthologs - mouse 79%, rat 83%.
This gene encodes a transmembrane protein which belongs to subfamily 35A of the solute carrier superfamily. The encoded protein is a nucleoside-sugar transporter. Alternative splicing results in multiple transcript variants.
Specifications
| SLC35A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC35A5 | |
| probable UDP-sugar transporter protein SLC35A5; SLC35A5; Solute carrier family 35 member A5; solute carrier family 35, member A5; UNQ164/PRO190 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55032 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BS91 | |
| SLC35A5 | |
| Recombinant Human SLC35A5. Recombinant protein control fragment (Product #RP-106805). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction