missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC34A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06366-50ul
This item is not returnable.
View return policy
Description
SLC34A3 Polyclonal antibody specifically detects SLC34A3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC34A3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Na(+)/Pi cotransporter 2C, Na(+)-dependent phosphate cotransporter 2C, naPi-2c, sodium-phosphate transport protein 2C, solute carrier family 34 (sodium phosphate), member 3 | |
| Produced in rabbits immunized with E. coli-derived Human SLC34A3 fragment. | |
| 50 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 142680 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction