missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC34A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£164.00 - £366.00
Specifications
| Antigen | SLC34A3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675880
|
Novus Biologicals
NBP2-93939-0.02ml |
0.02 mL |
£164.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647031
|
Novus Biologicals
NBP2-93939-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC34A3 Polyclonal antibody specifically detects SLC34A3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SLC34A3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 142680 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Na(+)/Pi cotransporter 2C, Na(+)-dependent phosphate cotransporter 2C, naPi-2c, sodium-phosphate transport protein 2C, solute carrier family 34 (sodium phosphate), member 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human SLC34A3 (NP_063949.2). MAAAAAAGSGTPREEEGPAGEAAASQPQAPTSVPGARLSRLPLARVKALVKADPDVTLAGQEAIFILARAAELFVETIAKDAYCCAQQGKRKTLQRRDLDNAIEAVDEFAFLEGTLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title