missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC34A2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21295-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC34A2 encodes a protein that is a pH-sensitive sodium-dependent phosphate transporter. Phosphate uptake is increased at lower pH. Defects in this gene are a cause of pulmonary alveolar microlithiasis. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| SLC34A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC34A2 | |
| Na(+)/Pi cotransporter 2B, NaPi 2b, NAPI 3B, NAPI IIb, NaPi3b, SLC34A2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10568 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95436 | |
| SLC34A2 | |
| SLC34A2 Fusion Protein Ag15846 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction