Learn More
SLC34A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562358
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PLPVPGGHVM RGTAFAYVPS PQVLHRIPGT SAYAFPSLGP VALAEHTCPC GEVLERHEPL PAKLALEEEQ KPESRLVPKL RQA Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
Renal tubular reabsorption of phosphate is critical to the maintenance of phosphate homeostasis in mammals. The brush-border membrane Na+/Pi cotransport systems in proximal tubules play a major role in this process. The renal Na(+)/P(i) cotransporter NPT2 is expressed in the brush border membrane (BBM) of proximal tubular cells. NPT2 gene expression is crucial for PTH effects on renal P(i) handling. However, renal expression of the sodium/phosphate cotransporter gene, NPT2, is not required for regulation of renal 1 alpha-hydroxylase by phosphate. NPT2 is an integral membrane protein expressed in kidney and lung. The gene encoding human NPT1 maps to chromosome 6q21.3-p23, while the gene encoding human NPT2 maps to chromosome 5q35.Specifications
SLC34A1 | |
Polyclonal | |
Unconjugated | |
SLC34A1 | |
FRTS2; HCINF2; Na(+)/Pi cotransporter 2A; Na(+)-dependent phosphate cotransporter 2A; Na/Pi cotransporter; Na+-phosphate cotransporter type II; NaPi-2; NaPi2A; naPi-2a; NAPI-3; naPi-7; NaPi-IIa; NPHLOP1; NPT2; Npt2a; NPTIIa; renal Na+/Pi transporter; renal sodium-dependent phosphate transporter; SLC11; Slc17a2; Slc34a1; sodium/phosphate co-transporter; sodium/phosphate cotransporter 2A; Sodium-dependent phosphate transport protein 2A; sodium-phosphate transport protein 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (sodium/hydrogen exchanger) member 2; solute carrier family 17 (sodium/hydrogen exchanger), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; solute carrier family 34 member 1; solute carrier family 34, member 1; type IIa Na+/Pi-cotransporter | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6569 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q06495 | |
SLC34A1 | |
Recombinant protein corresponding to Human SLC34A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only