missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC34A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577010
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Renal tubular reabsorption of phosphate is critical to the maintenance of phosphate homeostasis in mammals. The brush-border membrane Na+/Pi cotransport systems in proximal tubules play a major role in this process. The renal Na(+)/P(i) cotransporter NPT2 is expressed in the brush border membrane (BBM) of proximal tubular cells. NPT2 gene expression is crucial for PTH effects on renal P(i) handling. However, renal expression of the sodium/phosphate cotransporter gene, NPT2, is not required for regulation of renal 1 alpha-hydroxylase by phosphate. NPT2 is an integral membrane protein expressed in kidney and lung. The gene encoding human NPT1 maps to chromosome 6q21.3-p23, while the gene encoding human NPT2 maps to chromosome 5q35.
Specifications
| SLC34A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC34A1 | |
| FRTS2; HCINF2; Na(+)/Pi cotransporter 2A; Na(+)-dependent phosphate cotransporter 2A; Na/Pi cotransporter; Na+-phosphate cotransporter type II; NaPi-2; NaPi2A; naPi-2a; NAPI-3; naPi-7; NaPi-IIa; NPHLOP1; NPT2; Npt2a; NPTIIa; renal Na+/Pi transporter; renal sodium-dependent phosphate transporter; SLC11; Slc17a2; Slc34a1; sodium/phosphate co-transporter; sodium/phosphate cotransporter 2A; Sodium-dependent phosphate transport protein 2A; sodium-phosphate transport protein 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (sodium/hydrogen exchanger) member 2; solute carrier family 17 (sodium/hydrogen exchanger), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; solute carrier family 34 member 1; solute carrier family 34, member 1; type IIa Na+/Pi-cotransporter | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20505, 25548, 6569 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q06495, Q06496, Q60825 | |
| SLC34A1 | |
| Recombinant full length Human SLC34A1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction