missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC34A1 Antibody (10B1.3E9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £423.00
Specifications
| Antigen | SLC34A1 |
|---|---|
| Clone | 10B1.3E9 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50, Immunoprecipitation reported in scientific literature (PMID 35307350), Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606946
|
Novus Biologicals
NBP2-42216SS |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629796
|
Novus Biologicals
NBP2-42216 |
0.1 mg |
£423.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC34A1 Monoclonal antibody specifically detects SLC34A1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| SLC34A1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Endocrinology, Signal Transduction | |
| PBS | |
| FRTS2, Na(+)/Pi cotransporter 2A, Na(+)-dependent phosphate cotransporter 2A, NaPi-2a, NaPi-3, NPT2NPHLOP1, NPTIIa, renal sodium-dependent phosphate transporter, SLC11, SLC17A2Na+-phosphate cotransporter type II, sodium/phosphate co-transporter, Sodium/phosphate cotransporter 2A, sodium-dependent phosphate transport protein 2A, Sodium-phosphate transport protein 2A, solute carrier family 17 (sodium phosphate), member 2, solute carrier family 34 (sodium phosphate), member 1, Solute carrier family 34 member 1 | |
| Partial synthetic peptide made to an internal portion of the human SLC34A1 protein (between amino acids 25-150) [UniProt Q06495] | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 10B1.3E9 | |
| Western Blot 1:500, Flow Cytometry 1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50, Immunoprecipitation reported in scientific literature (PMID 35307350), Immunohistochemistry-Paraffin 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Q06495 | |
| 6569 | |
| IgG1 κ | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title