missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC33A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106589
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC33A1.
SLC33A1 is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm.The encoded protein is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm.
Specifications
| SLC33A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC33A1 | |
| Acatn; acetyl-CoA transporter 1; acetyl-coenzyme A transporter; acetyl-coenzyme A transporter 1; AI315656; AI788741; AT1; AT-1; CCHLND; D630022N01Rik; similar to solute carrier family 33 (acetyl-CoA transporter), member 1; SLC33A1; solute carrier family 33 (acetyl-CoA transporter), member 1; solute carrier family 33 member 1; SPG42; zgc:63693 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11416, 9197 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00400, Q99J27 | |
| SLC33A1 | |
| A synthesized peptide derived from human SLC33A1(Accession O00400), corresponding to amino acid residues L189-D239. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction