missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC31A1/CTR1 Rabbit anti-Human, Mouse, Rat, Clone: 4C10C5, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | SLC31A1/CTR1 |
|---|---|
| Clone | 4C10C5 |
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
18315644
|
Novus Biologicals
NBP3-15304-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18335303
|
Bio-Techne
NBP3-15304-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human, Mouse, Rat samples. It is validated for Western BlotEspecificaciones
| SLC31A1/CTR1 | |
| Western Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC31A1/CTR1 (O15431). MDHSHHMGMSYMDSNSTMQPSHHHPTTSASHSHGGGDSSMMMMPMTFYFGFKNVELLFSGLVINTAGEMAGAFVAVFLLAMFYEGLKIARESLLRKSQVS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4C10C5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Neurotransmission, Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1317 | |
| IgG | |
| Affinity purified |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto