missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101996
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total SLC30A9.
The gene corresponding to embryonic lung protein [also known as Solute carrier family 30 (Zinc transporter), member 9, SLC30A9] is likely to be an evolutionarily conserved, housekeeping gene that plays a role intimately linked with cellular replication, DNA synthesis and/or transcriptional regulation.
Spécification
| SLC30A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A9 | |
| 2310024J23Rik; AL024256; C4orf1; expressed in human embryonic lung; GAC63; GRIP1-associated coactivator 1; GRIP1-associated coactivator 63; GRIP1-dependent nuclear receptor coactivator; HUEL; Human embryonic lung protein; Slc30a9; solute carrier family 30 (zinc transporter), member 9; solute carrier family 30 member 9; Zinc transporter 9; ZNT9; znT-9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10463, 109108, 498358 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5IRJ6, Q6PML9 | |
| SLC30A9 | |
| A synthesized peptide derived from human SLC30A9(Accession Q6PML9), corresponding to amino acid residues S171-N221. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu