Learn More
Invitrogen™ SLC30A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588194
Description
Immunogen sequence: KVLLQTTPPH VIGQLDKLIR EVSTLDGVLE VRNEHFWTLG FGSLAGSVHV RIRRDANEQM VLAHVTNRLY TLVSTLTVQI FKDDWIRPAL LSGPVAANVL NFSDHHVIPM PLLKGTDDLN PVTSTPAKPS SPPPEFSFNT PGKNVNPVIL LNTQTRPYGF GLNHGHTPYS SMLNQGLGVP GIGATQGLRT GFTNIPSRYG TNNRIGQPRP; Positive Samples: U-87MG, Mouse brain, Rat brain; Cellular Location: Golgi apparatus, Multi-pass membrane protein, trans-Golgi network membrane.
Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
Specifications
| SLC30A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A6 | |
| 9530029F08Rik; MST103; MSTP103; SLC30A6; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 6; solute carrier family 30 member 6; Zinc transporter 6; zinc transporter-like 3; Znt6; znT-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 210148, 298786, 55676 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.11 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6NXT4, Q8BJM5 | |
| SLC30A6 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.