missing translation for 'onlineSavingsMsg'
Learn More

SLC30A6 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £366.00

Specifications

Antigen SLC30A6
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18669240
View Documents
Novus Biologicals
NBP2-93596-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18653750
View Documents
Novus Biologicals
NBP2-93596-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC30A6 Polyclonal antibody specifically detects SLC30A6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

SLC30A6
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
MGC45055, MST103, MSTP103, solute carrier family 30 (zinc transporter), member 6, Solute carrier family 30 member 6, zinc transporter 6, ZnT-6, ZNT6FLJ31101
Recombinant fusion protein containing a sequence corresponding to amino acids 252-461 of human SLC30A6 (NP_060434.2). KVLLQTTPPHVIGQLDKLIREVSTLDGVLEVRNEHFWTLGFGSLAGSVHVRIRRDANEQMVLAHVTNRLYTLVSTLTVQIFKDDWIRPALLSGPVAANVLNFSDHHVIPMPLLKGTDDLNPVTSTPAKPSSPPPEFSFNTPGKNVNPVILLNTQTRPYGFGLNHGHTPYSSMLNQGLGVPGIGATQGLRTGFTNIPSRYGTNNRIGQPRP
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
55676
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.