missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553749
This item is not returnable.
View return policy
Description
Immunogen sequence: PSHLNVDYIK EALMKIEDVY SVEDLNIWSL TSGKSTAIVH IQLIPGSSSK WEEVQSKANH LLLNTFGMYR CTIQLQSYRQ EVDRTCANCQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Zinc is the second most abundant trace metal in the human body. It is an essential element, serving both a structural role, as in the formation of zinc fingers in DNA-binding proteins, and a catalytic role in metalloenzymes, such as pancreatic carboxypeptidases (e.g., MIM 114852), alkaline phosphatases (e.g., MIM 171760), various dehydrogenases, and superoxide dismutases (e.g., MIM 147450). SLC30A4, or ZNT4, belongs to the ZNT family of zinc transporters. ZNTs are involved in transporting zinc out of the cytoplasm and have similar structures, consisting of 6 transmembrane domains and a histidine-rich cytoplasmic loop (Huang and Gitschier, 1997).
Specifications
| SLC30A4 | |
| Polyclonal | |
| Unconjugated | |
| Slc30a4 | |
| dri 27 protein; Dri 27/ZnT4 protein; lethal milk protein; Lm; Slc30a4; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 4; zinc transporter 4; solute carrier family 30 member 4; Zinc transporter 4; ZNT4; znT-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14863 | |
| Slc30a4 | |
| Recombinant protein corresponding to Human SLC30A4. Recombinant protein control fragment (Product #RP-91249). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction