missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569263
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
Slc30a3 encodes a protein that is involved in accumulation of zinc in synaptic vesicles.
Specifications
| SLC30A3 | |
| Polyclonal | |
| Unconjugated | |
| Slc30a3 | |
| SLC30A3; solute carrier family 30 (zinc transporter), member 3; solute carrier family 30 member 3; solute carrier family 30 protein; zinc transporter 3; zinc transporter ZnT-3; ZNT3; znT-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7781 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99726 | |
| Slc30a3 | |
| synthetic peptide directed towards the middle region of human SLC30A3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction