missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67993-1-IG-20UL
This item is not returnable.
View return policy
Description
The SLC30A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC30A2. This antibody recognizes human, mouse antigen. The SLC30A2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| SLC30A2 | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ36708; MGC11303; PP12488; ZNT2; ZnT-2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 41 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D9E5 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7780 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction