missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537463
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 55 kDa. Purity is >95% by SDS-PAGE.
Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
Specifications
| SLC30A1 | |
| Polyclonal | |
| Unconjugated | |
| Slc30a1 | |
| AI839647; C130040I11Rik; Slc30a1; solute carrier family 30 (zinc transporter), member 1; solute carrier family 30 member 1; solute carrier family 30, member 1; zinc transporter 1; ZNT1; ZnT-1; ZnT-1 zinc transporter; ZRC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22782, 58976, 7779 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q60738, Q62720, Q9Y6M5 | |
| Slc30a1 | |
| Synthetic peptide corresponding to amino acids 200-250 of human ZnT-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction