missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577768
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Per vendor datasheet, reconstitute with 50uL deionized water.
Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
Specifications
| SLC30A1 | |
| Polyclonal | |
| Unconjugated | |
| Slc30a1 | |
| SLC30A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22782, 58976 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q60738, Q62720 | |
| Slc30a1 | |
| Peptide (C)ENLEKKPRRTKAE, corresponding to amino acid residues 473- 485 of rat ZnT1 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction