Learn More
SLC2A9 Mouse anti-Human, Unconjugated, Clone: 2D6E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67530-1-IG-20UL
Description
The SLC2A9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC2A9. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SLC2A9 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| SLC2A9 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2D6E1 | |
| Unconjugated | |
| 20 μL | |
| 56606 | |
| IgG1 |