Learn More
Invitrogen™ SLC2A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595120
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: U87 Cell, 293T Cell, HT1080 Cell, SW620 Cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
Specifications
| SLC2A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A5 | |
| AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6518 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P22732 | |
| SLC2A5 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human SLC2A5 (481-501aa VSEVYPEKEELKELPPVTSEQ). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.