missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC2A12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568915
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Goat: 86%; Horse: 86%; Human: 86%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 93%.
Solute carrier family 2, facilitated glucose transporter member 12 is a protein that in humans is encoded by the SLC2A12 gene. SLC2A12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. This family of transporters show conservation of 12 transmembrane helices as well as functionally significant amino acid residues.
Specifications
| SLC2A12 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A12 | |
| facilitated glucose transporter; glucose transporter type 12; GLUT12; GLUT-12; GLUT8; SLC2A12; solute carrier family 2 (facilitated glucose transporter), member 12; solute carrier family 2 member 12; solute carrier family 2, facilitated glucose transporter member 12; solute carrier family 2, member 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 353169 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BFW9 | |
| SLC2A12 | |
| synthetic peptide corresponding to a region of mouse SLC2A12 (aa 551-600). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction