Learn More
SLC29A4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26423-1-AP-150UL
Description
The SLC29A4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC29A4. This antibody recognizes human, rat antigen. The SLC29A4 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| SLC29A4 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| ENT4, hENT4, PMAT, SLC29A4 | |
| 150 μL | |
| 222962 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |