missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC29A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£484.00
Specifications
| Antigen | SLC29A4 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC29A4 Polyclonal specifically detects SLC29A4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| SLC29A4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| ENT4PMAT, equilibrative nucleoside transporter 4, FLJ34923, hENT4, Plasma membrane monoamine transporter, solute carrier family 29 (nucleoside transporters), member 4, Solute carrier family 29 member 4 | |
| SLC29A4 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 222962 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QGQGLRARGVPAFTDTTLDEPVPDDRY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title